siRNA Details
Gene Symbol: MTFR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099625.1 | NP_001093095.1 | 6 | 763 | ||
NM_001099626.1 | NP_001093096.1 | 6 | 815 | |||
NM_019557.5 | NP_062457.3 | 6 | 861 | |||
GenBank mRNA | AB075880.1 | - | 6 | 691 | ||
AF173891.1 | - | 6 | 628 | |||
AK127164.1 | - | 2 | 1297 | |||
AK299221.1 | - | 6 | 706 | |||
AK314297.1 | - | 6 | 691 | |||
AL512766.1 | - | 6 | 863 | |||
AY007100.1 | - | 6 | 741 | |||
BC017175.2 | - | 6 | 815 | 3162883 | ||
KU178723.1 | - | 5 | 553 | |||
KU178724.1 | - | 5 | 576 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
