siRNA Details
Gene Symbol: KIAA1217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019590.4 | NP_062536.2 | 19 | 5343 | ||
XM_005252516.3 | XP_005252573.1 | 14 | 4249 | |||
XM_011519552.2 | XP_011517854.2 | 20 | 5398 | |||
XM_011519555.2 | XP_011517857.2 | 19 | 5316 | |||
XM_011519558.2 | XP_011517860.2 | 19 | 5280 | |||
XM_011519559.2 | XP_011517861.2 | 18 | 4985 | |||
XM_017016416.1 | XP_016871905.1 | 20 | 5424 | |||
XM_017016417.1 | XP_016871906.1 | 18 | 5238 | |||
XM_017016418.1 | XP_016871907.1 | 18 | 6060 | |||
XM_017016419.1 | XP_016871908.1 | 17 | 5955 | |||
XM_017016420.1 | XP_016871909.1 | 18 | 4807 | |||
XM_017016421.1 | XP_016871910.1 | 20 | 5017 | |||
XM_017016422.1 | XP_016871911.1 | 17 | 4702 | |||
XM_017016423.1 | XP_016871912.1 | 15 | 4354 | |||
XM_017016424.1 | XP_016871913.1 | 15 | 4426 | |||
GenBank mRNA | AK055179.1 | - | 1 | 679 | ||
AK125675.1 | - | 1 | 1200 | |||
AL833280.1 | - | 19 | 4971 | |||
BX640796.1 | - | 14 | 4250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
