siRNA Details
Gene Symbol: KYAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008661.2 | NP_001008661.1 | 12, 13 | 1584 | ||
NM_001008662.2 | NP_001008662.1 | 11, 12 | 1484 | |||
GenBank mRNA | AF091090.1 | - | 8, 9 | 787 | ||
AK057176.1 | - | 11, 12 | 1300 | |||
AK301326.1 | - | 12, 13 | 1271 | |||
AK315917.1 | - | 12, 13 | 1281 | |||
AL833593.1 | - | 13, 14 | 1386 | |||
AY028624.1 | - | 12, 13 | 1404 | |||
BC000819.1 | - | 9, 10 | 957 | 3453829 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
