siRNA Details
Gene Symbol: PANX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160300.1 | NP_001153772.1 | 3 | 1849 | ||
NM_052839.3 | NP_443071.2 | 3 | 1849 | |||
NR_027691.1 | 4 | 1900 | ||||
GenBank mRNA | AF398510.1 | - | 3 | 1819 | ||
AF398511.1 | - | 4 | 1870 | |||
AK096019.1 | - | 3 | 1833 | |||
AK299910.1 | - | 3 | 1849 | |||
BC101023.2 | - | 6 | 1728 | 40007400 | ||
CR456467.1 | - | 2 | 1499 |

More Information
Other Species Targeted

Gene Ontology Categories:
