siRNA Details
Gene Symbol: PANX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160300.1 | NP_001153772.1 | 3, 4 | 1889 | ||
NR_027691.1 | 4, 5 | 1940 | ||||
GenBank mRNA | AF398510.1 | - | 3, 4 | 1859 | ||
AF398511.1 | - | 4, 5 | 1910 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
