siRNA Details
Gene Symbol: PANX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160300.1 | NP_001153772.1 | 1, 2 | 220 | ||
NM_052839.3 | NP_443071.2 | 1, 2 | 220 | |||
GenBank mRNA | AF398510.1 | - | 1, 2 | 190 | ||
AK096019.1 | - | 1, 2 | 204 | |||
AK299910.1 | - | 1, 2 | 220 | |||
DA116770.1 | - | 1, 2 | 222 | |||
DA127683.1 | - | 1, 2 | 220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
