siRNA Details
Gene Symbol: CNGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286130.1 | NP_001273059.1 | 32 | 3390 | ||
NM_001297.4 | NP_001288.3 | 32 | 3408 | |||
XM_011522870.2 | XP_011521172.1 | 20 | 2677 | |||
GenBank mRNA | AF042498.1 | - | 32 | 3395 | ||
U58837.1 | - | 32 | 3400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
