siRNA Details
Gene Symbol: CPA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020361.4 | NP_065094.3 | 3 | 479 | ||
XM_017013647.1 | XP_016869136.1 | 3 | 1208 | |||
GenBank mRNA | AF221594.1 | - | 3 | 436 | ||
BC033684.1 | - | 3 | 439 | 5193403 | ||
BI756197.1 | - | 3 | 479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
