siRNA Details
Gene Symbol: CPA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020361.4 | NP_065094.3 | 1, 2 | 380 | ||
XM_011517570.2 | XP_011515872.1 | 1, 2 | 919 | |||
XM_017013647.1 | XP_016869136.1 | 1, 2 | 1109 | |||
GenBank mRNA | AF221594.1 | - | 1, 2 | 337 | ||
AY044833.1 | - | 1, 2 | 338 | |||
BC033684.1 | - | 1, 2 | 340 | 5193403 | ||
BI756197.1 | - | 1, 2 | 380 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
