siRNA Details
Gene Symbol: RAB25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020387.3 | NP_065120.2 | 3, 4 | 678 | ||
NR_133653.1 | 2, 3 | 482 | ||||
GenBank mRNA | AF083124.1 | - | 3, 4 | 639 | ||
AF274025.1 | - | 3, 4 | 657 | |||
BC004416.1 | - | 1, 2 | 168 | |||
BC009831.2 | - | 3, 4 | 652 | 3926839 | ||
BC033322.1 | - | 3, 4 | 642 | 4775725 | ||
BP273992.1 | - | 2, 3 | 467 |
