siRNA Details
Gene Symbol: KIAA1147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080392.1 | NP_001073861.1 | 5 | 709 | ||
GenBank mRNA | AB032973.2 | - | 5 | 698 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KIAA1147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080392.1 | NP_001073861.1 | 5 | 709 | ||
GenBank mRNA | AB032973.2 | - | 5 | 698 |