siRNA Details
Gene Symbol: ZNF248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267597.1 | NP_001254526.1 | 6 | 665 | ||
NM_021045.2 | NP_066383.1 | 6 | 1037 | |||
XM_006717485.3 | XP_006717548.1 | 5 | 994 | |||
XM_011519597.2 | XP_011517899.1 | 5 | 586 | |||
XM_017016444.1 | XP_016871933.1 | 4 | 643 | |||
XM_017016445.1 | XP_016871934.1 | 5 | 1134 | |||
XM_017016446.1 | XP_016871935.1 | 6 | 685 | |||
GenBank mRNA | AJ491695.1 | - | 6 | 959 | ||
AK299357.1 | - | 6 | 665 | |||
BC132915.1 | - | 6 | 695 | 40146937 | ||
BC132917.1 | - | 6 | 695 | 40146939 |
