siRNA Details
Gene Symbol: ZNF287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020653.2 | NP_065704.2 | 5 | 1131 | ||
XM_011523968.2 | XP_011522270.1 | 5 | 1577 | |||
XM_017024887.1 | XP_016880376.1 | 5 | 1338 | |||
GenBank mRNA | AF217227.1 | - | 5 | 1075 | ||
BC032002.1 | - | 4 | 751 | |||
BC041159.1 | - | 5 | 1296 | |||
BC057791.1 | - | 5 | 1131 | |||
BC101487.1 | - | 4 | 750 | 8068993 | ||
BC113363.1 | - | 4 | 754 | 8322415 | ||
CR457194.1 | - | 4 | 657 |
