siRNA Details
Gene Symbol: KIAA1257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020741.2 | NP_065792.1 | 6 | 1080 | ||
XM_011513050.2 | XP_011511352.2 | 6 | 980 | |||
XM_017006939.1 | XP_016862428.1 | 7 | 1535 | |||
XM_017006940.1 | XP_016862429.1 | 7 | 1531 | |||
XM_017006941.1 | XP_016862430.1 | 7 | 1108 | |||
XM_017006942.1 | XP_016862431.1 | 7 | 1502 | |||
XM_017006943.1 | XP_016862432.1 | 7 | 1260 | |||
XM_017006944.1 | XP_016862433.1 | 6 | 1080 | |||
XM_017006945.1 | XP_016862434.1 | 6 | 980 | |||
XM_017006946.1 | XP_016862435.1 | 5 | 691 | |||
XM_017006947.1 | XP_016862436.1 | 4 | 575 | |||
XM_017006948.1 | XP_016862437.1 | 6 | 752 | |||
XM_017006949.1 | XP_016862438.1 | 5 | 699 | |||
XM_017006950.1 | XP_016862439.1 | 5 | 6186 | |||
XM_017006951.1 | XP_016862440.1 | 5 | 6291 | |||
XM_017006952.1 | XP_016862441.1 | 4 | 469 | |||
XM_017006953.1 | XP_016862442.1 | 4 | 475 | |||
GenBank mRNA | AB033083.1 | - | 6 | 1052 | ||
BC031632.1 | - | 5 | 1041 | 5170293 | ||
BC038736.1 | - | 6 | 1013 |
Gene Symbol: LOC100132731 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056377.1 | - | 5 | 1656 |
