siRNA Details
Gene Symbol: KIAA1257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020741.2 | NP_065792.1 | 3 | 578 | ||
XM_011513050.2 | XP_011511352.2 | 3 | 478 | |||
XM_017006939.1 | XP_016862428.1 | 4 | 1033 | |||
XM_017006940.1 | XP_016862429.1 | 4 | 1029 | |||
XM_017006941.1 | XP_016862430.1 | 4 | 606 | |||
XM_017006942.1 | XP_016862431.1 | 4 | 1000 | |||
XM_017006943.1 | XP_016862432.1 | 4 | 758 | |||
XM_017006944.1 | XP_016862433.1 | 3 | 578 | |||
XM_017006945.1 | XP_016862434.1 | 3 | 478 | |||
XM_017006946.1 | XP_016862435.1 | 2 | 189 | |||
XM_017006948.1 | XP_016862437.1 | 3 | 250 | |||
XM_017006949.1 | XP_016862438.1 | 2 | 197 | |||
XM_017006950.1 | XP_016862439.1 | 2 | 5684 | |||
GenBank mRNA | AB033083.1 | - | 3 | 550 | ||
BC031632.1 | - | 2 | 539 | 5170293 | ||
BC038736.1 | - | 3 | 511 |
Gene Symbol: LOC100132731 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056377.1 | - | 2 | 1154 |
