siRNA Details
Gene Symbol: NUFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020772.2 | NP_065823.1 | 2 | 745 | ||
XM_017024896.1 | XP_016880385.1 | 2 | 2043 | |||
GenBank mRNA | AB037742.1 | - | 2 | 715 | ||
AJ493465.1 | - | 2 | 656 | |||
AK293075.1 | - | 2 | 745 | |||
AY232289.1 | - | 2 | 724 | |||
BC108307.1 | - | 2 | 702 | 6502841 | ||
BC129990.1 | - | 2 | 660 | 40128910 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
