siRNA Details
Gene Symbol: NUFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB037742.1 | - | 2 | 1385 | ||
AJ493465.1 | - | 2 | 1326 | |||
BC108307.1 | - | 2 | 1372 | 6502841 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
