siRNA Details
Gene Symbol: NUFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020772.2 | NP_065823.1 | 1, 2 | 372 | ||
GenBank mRNA | AB037742.1 | - | 1, 2 | 342 | ||
AJ493465.1 | - | 1, 2 | 283 | |||
AK293075.1 | - | 1, 2 | 372 | |||
AY232289.1 | - | 1, 2 | 351 | |||
BC108307.1 | - | 1, 2 | 329 | 6502841 | ||
BC129990.1 | - | 1, 2 | 287 | 40128910 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
