siRNA Details
Gene Symbol: TBC1D14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113361.1 | NP_001106832.1 | 8 | 1418 | ||
NM_001113363.1 | NP_001106834.1 | 6 | 684 | |||
NM_001286805.1 | NP_001273734.1 | 6 | 553 | |||
NM_020773.2 | NP_065824.2 | 8 | 1540 | |||
XM_005247986.2 | XP_005248043.1 | 8 | 1531 | |||
XM_006713895.2 | XP_006713958.1 | 8 | 1708 | |||
XM_011513507.2 | XP_011511809.1 | 8 | 1525 | |||
XM_011513508.1 | XP_011511810.1 | 8 | 1418 | |||
XM_011513513.2 | XP_011511815.1 | 6 | 620 | |||
XM_017008477.1 | XP_016863966.1 | 8 | 1418 | |||
XM_017008478.1 | XP_016863967.1 | 7 | 790 | |||
XM_017008479.1 | XP_016863968.1 | 6 | 626 | |||
XM_017008480.1 | XP_016863969.1 | 8 | 750 | |||
GenBank mRNA | AB037743.1 | - | 7 | 1368 | ||
AB449900.1 | - | 7 | 1339 | |||
AK056962.1 | - | 3 | 790 | |||
AK295068.1 | - | 3 | 196 | |||
AL833868.1 | - | 5 | 405 | |||
BC041167.1 | - | 8 | 1463 | 5575580 | ||
BQ230652.1 | - | 6 | 684 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
