siRNA Details
Gene Symbol: TBC1D14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113361.1 | NP_001106832.1 | 2, 3 | 809 | ||
NM_020773.2 | NP_065824.2 | 2, 3 | 931 | |||
XM_005247986.2 | XP_005248043.1 | 2, 3 | 922 | |||
XM_006713895.2 | XP_006713958.1 | 2, 3 | 1099 | |||
XM_011513507.2 | XP_011511809.1 | 2, 3 | 916 | |||
XM_011513508.1 | XP_011511810.1 | 2, 3 | 809 | |||
XM_017008477.1 | XP_016863966.1 | 2, 3 | 809 | |||
GenBank mRNA | AB037743.1 | - | 1, 2 | 759 | ||
AB449900.1 | - | 1, 2 | 730 | |||
BC041167.1 | - | 2, 3 | 854 | 5575580 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
