siRNA Details
Gene Symbol: KIAA1324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267048.1 | NP_001253977.1 | 18 | 2665 | ||
NM_001284352.1 | NP_001271281.1 | 19 | 2805 | |||
NM_001284353.1 | NP_001271282.1 | 13 | 2786 | |||
NM_020775.4 | NP_065826.2 | 20 | 2926 | |||
XM_011541825.1 | XP_011540127.1 | 20 | 2786 | |||
XM_011541826.2 | XP_011540128.1 | 20 | 2786 | |||
XM_011541827.1 | XP_011540129.1 | 19 | 2636 | |||
XM_017001883.1 | XP_016857372.1 | 19 | 2784 | |||
GenBank mRNA | AB037745.1 | - | 11 | 1434 | ||
AK026832.1 | - | 12 | 1560 | |||
AK057647.1 | - | 19 | 2634 | |||
AK122934.1 | - | 13 | 2786 | |||
AK297221.1 | - | 19 | 2599 | |||
AK301598.1 | - | 17 | 2141 | |||
AY358366.1 | - | 21 | 2842 | |||
BC031648.1 | - | 12 | 1862 | |||
BC125208.1 | - | 18 | 2473 | 40123480 |
