siRNA Details
Gene Symbol: KIAA1324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267048.1 | NP_001253977.1 | 11 | 1573 | ||
NM_001284352.1 | NP_001271281.1 | 12 | 1713 | |||
NM_001284353.1 | NP_001271282.1 | 6 | 1694 | |||
NM_020775.4 | NP_065826.2 | 13 | 1834 | |||
XM_011541825.1 | XP_011540127.1 | 13 | 1694 | |||
XM_011541826.2 | XP_011540128.1 | 13 | 1694 | |||
XM_011541827.1 | XP_011540129.1 | 12 | 1544 | |||
XM_017001883.1 | XP_016857372.1 | 12 | 1692 | |||
GenBank mRNA | AB037745.1 | - | 4 | 342 | ||
AK026832.1 | - | 5 | 468 | |||
AK057647.1 | - | 12 | 1542 | |||
AK122934.1 | - | 6 | 1694 | |||
AK297221.1 | - | 12 | 1507 | |||
AK297478.1 | - | 12 | 1436 | |||
AK301598.1 | - | 10 | 1049 | |||
AY358366.1 | - | 14 | 1750 | |||
BC031648.1 | - | 5 | 770 | |||
BC125208.1 | - | 11 | 1381 | 40123480 |
