siRNA Details
Gene Symbol: ARRDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020801.2 | NP_065852.1 | 5 | 1098 | ||
GenBank mRNA | AB037797.1 | - | 5 | 1070 | ||
AK291753.1 | - | 5 | 1073 | |||
BC015928.2 | - | 5 | 1065 | 3908916 | ||
BC053619.1 | - | 5 | 1070 | 6155815 |
