siRNA Details
Gene Symbol: RIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135920.2 | NP_001129392.2 | 18 | 2284 | ||
NM_001206557.1 | NP_001193486.1 | 17 | 2173 | |||
NM_020829.3 | NP_065880.2 | 18 | 2284 | |||
XM_005251523.3 | XP_005251580.1 | 18 | 2370 | |||
XM_011517967.2 | XP_011516269.1 | 11 | 1282 | |||
XM_011517968.2 | XP_011516270.1 | 6 | 617 | |||
XM_017014934.1 | XP_016870423.1 | 18 | 1940 | |||
XM_017014935.1 | XP_016870424.1 | 16 | 2841 | |||
XM_017014936.1 | XP_016870425.1 | 16 | 1780 | |||
GenBank mRNA | AB037853.2 | - | 2 | 212 | ||
AB205401.1 | - | 17 | 1878 | |||
AK022642.1 | - | 4 | 504 | |||
AK074150.1 | - | 2 | 130 | |||
AK124394.1 | - | 14 | 2792 | |||
AL136875.1 | - | 18 | 2076 | |||
BC136616.1 | - | 17 | 1878 | 9020606 | ||
BC144296.1 | - | 16 | 1767 | 9052816 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
