siRNA Details
Gene Symbol: RIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206557.1 | NP_001193486.1 | 25 | 4269 | ||
NM_020829.3 | NP_065880.2 | 26 | 4380 | |||
XM_005251523.3 | XP_005251580.1 | 26 | 4466 | |||
XM_011517967.2 | XP_011516269.1 | 19 | 3378 | |||
XM_011517968.2 | XP_011516270.1 | 14 | 2713 | |||
XM_017014934.1 | XP_016870423.1 | 26 | 4036 | |||
XM_017014935.1 | XP_016870424.1 | 24 | 4937 | |||
XM_017014936.1 | XP_016870425.1 | 24 | 3876 | |||
GenBank mRNA | AB037853.2 | - | 10 | 2308 | ||
AB205401.1 | - | 25 | 3974 | |||
AK074150.1 | - | 10 | 2226 | |||
BC030576.1 | - | 6 | 1157 | |||
BC136616.1 | - | 25 | 3974 | 9020606 | ||
BC144296.1 | - | 24 | 3863 | 9052816 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
