siRNA Details
Gene Symbol: RIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135920.2 | NP_001129392.2 | 21 | 3234 | ||
NM_001206557.1 | NP_001193486.1 | 20 | 3123 | |||
NM_020829.3 | NP_065880.2 | 21 | 3234 | |||
XM_005251523.3 | XP_005251580.1 | 21 | 3320 | |||
XM_011517967.2 | XP_011516269.1 | 14 | 2232 | |||
XM_011517968.2 | XP_011516270.1 | 9 | 1567 | |||
XM_017014934.1 | XP_016870423.1 | 21 | 2890 | |||
XM_017014935.1 | XP_016870424.1 | 19 | 3791 | |||
XM_017014936.1 | XP_016870425.1 | 19 | 2730 | |||
GenBank mRNA | AB037853.2 | - | 5 | 1162 | ||
AB205401.1 | - | 20 | 2828 | |||
AK022642.1 | - | 7 | 1454 | |||
AK074150.1 | - | 5 | 1080 | |||
AK124394.1 | - | 17 | 3742 | |||
AL136875.1 | - | 21 | 3026 | |||
BC136616.1 | - | 20 | 2828 | 9020606 | ||
BC144296.1 | - | 19 | 2717 | 9052816 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
