siRNA Details
Gene Symbol: EBF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110514.1 | NP_001103984.1 | 12 | 1293 | ||
XM_006723600.1 | XP_006723663.1 | 12 | 1088 | |||
XM_017027983.1 | XP_016883472.1 | 13 | 1185 | |||
XM_017027984.1 | XP_016883473.1 | 13 | 1291 | |||
XM_017027985.1 | XP_016883474.1 | 6 | 447 | |||
GenBank mRNA | AB037863.1 | - | 12 | 1287 | ||
BC037555.1 | - | 4 | 393 |
