siRNA Details
Gene Symbol: ZBTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020861.2 | NP_065912.1 | 3 | 527 | ||
XM_005267076.2 | XP_005267133.1 | 3 | 388 | |||
XM_011536004.2 | XP_011534306.1 | 3 | 609 | |||
XM_017011137.1 | XP_016866626.1 | 4 | 787 | |||
GenBank mRNA | AK092886.1 | - | 3 | 379 | ||
AK291942.1 | - | 3 | 470 | |||
AL832957.1 | - | 1 | 299 | |||
BC020172.1 | - | 3 | 369 | 4577244 | ||
DB215154.1 | - | 3 | 528 |
