siRNA Details
Gene Symbol: NYAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020864.1 | NP_065915.1 | 2, 3 | 402 | ||
XM_005246708.3 | XP_005246765.1 | 3, 4 | 1273 | |||
XM_011511524.1 | XP_011509826.1 | 3, 4 | 1273 | |||
XM_017004564.1 | XP_016860053.1 | 2, 3 | 826 | |||
GenBank mRNA | AB040919.1 | - | 2, 3 | 310 |
