siRNA Details
Gene Symbol: ALS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020919.3 | NP_065970.2 | 13 | 2891 | ||
XM_006712654.2 | XP_006712717.1 | 13 | 3468 | |||
XM_006712655.3 | XP_006712718.1 | 9 | 1105 | |||
XM_011511530.2 | XP_011509832.1 | 12 | 2603 | |||
XM_017004569.1 | XP_016860058.1 | 12 | 2595 | |||
XM_017004570.1 | XP_016860059.1 | 12 | 2595 | |||
XM_017004571.1 | XP_016860060.1 | 12 | 2533 | |||
XM_017004572.1 | XP_016860061.1 | 10 | 2183 | |||
GenBank mRNA | AB046783.2 | - | 12 | 2612 | ||
AB053305.1 | - | 13 | 2658 | |||
AF391100.1 | - | 13 | 2658 | |||
AK056413.1 | - | 4 | 496 | |||
AK291029.1 | - | 13 | 2675 | |||
BC071576.1 | - | 13 | 2653 | 30347073 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
