siRNA Details
Gene Symbol: ALS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020919.3 | NP_065970.2 | 28, 29 | 4766 | ||
XM_006712654.2 | XP_006712717.1 | 28, 29 | 5343 | |||
XM_006712655.3 | XP_006712718.1 | 24, 25 | 2980 | |||
XM_011511530.2 | XP_011509832.1 | 27, 28 | 4478 | |||
XM_017004569.1 | XP_016860058.1 | 27, 28 | 4467 | |||
XM_017004571.1 | XP_016860060.1 | 27, 28 | 4405 | |||
XM_017004572.1 | XP_016860061.1 | 25, 26 | 4058 | |||
GenBank mRNA | AB046783.2 | - | 27, 28 | 4487 | ||
AB053305.1 | - | 28, 29 | 4533 | |||
AF391100.1 | - | 28, 29 | 4533 | |||
AK056413.1 | - | 20, 21 | 2514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
