siRNA Details
Gene Symbol: ALS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020919.3 | NP_065970.2 | 29 | 4849 | ||
XM_006712654.2 | XP_006712717.1 | 29 | 5426 | |||
XM_006712655.3 | XP_006712718.1 | 25 | 3063 | |||
XM_011511530.2 | XP_011509832.1 | 28 | 4561 | |||
XM_017004569.1 | XP_016860058.1 | 28 | 4550 | |||
XM_017004571.1 | XP_016860060.1 | 28 | 4488 | |||
XM_017004572.1 | XP_016860061.1 | 26 | 4141 | |||
GenBank mRNA | AB046783.2 | - | 28 | 4570 | ||
AB053305.1 | - | 29 | 4616 | |||
AF391100.1 | - | 29 | 4616 | |||
AK056413.1 | - | 21 | 2597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
