siRNA Details
Gene Symbol: FAM160B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135051.1 | NP_001128523.1 | 7 | 1320 | ||
NM_020940.3 | NP_065991.3 | 7 | 1320 | |||
GenBank mRNA | AB046820.2 | - | 6 | 915 | ||
BC037207.1 | - | 7 | 1320 | 4828347 | ||
CR933692.1 | - | 7 | 1040 |
