siRNA Details
Gene Symbol: GBA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277225.1 | NP_001264154.1 | 3 | 941 | ||
NM_020973.4 | NP_066024.1 | 3 | 1039 | |||
NR_102355.1 | 3 | 1039 | ||||
NR_102357.1 | 3 | 941 | ||||
GenBank mRNA | AB017913.1 | - | 3 | 988 | ||
AF317840.1 | - | 3 | 937 | |||
AF323990.1 | - | 3 | 986 | |||
AJ278964.1 | - | 3 | 937 | |||
AK292746.1 | - | 3 | 986 | |||
AK298377.1 | - | 3 | 941 | |||
AK313189.1 | - | 3 | 991 | |||
BC101829.1 | - | 3 | 974 | 8069335 | ||
BC136264.1 | - | 3 | 974 | 9020251 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
