siRNA Details
Gene Symbol: CATSPERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021185.4 | NP_067008.3 | 20, 21 | 2569 | ||
XM_005259114.4 | XP_005259171.1 | 20, 21 | 2665 | |||
XM_005259115.4 | XP_005259172.1 | 20, 21 | 2588 | |||
XM_005259116.2 | XP_005259173.1 | 18, 19 | 2389 | |||
XM_006723308.3 | XP_006723371.1 | 20, 21 | 3185 | |||
XM_006723310.2 | XP_006723373.1 | 20, 21 | 2452 | |||
XM_006723311.2 | XP_006723374.1 | 20, 21 | 2590 | |||
XM_011527174.2 | XP_011525476.1 | 20, 21 | 2579 | |||
XM_011527175.2 | XP_011525477.1 | 20, 21 | 2485 | |||
XM_011527176.2 | XP_011525478.1 | 20, 21 | 2488 | |||
XM_011527177.2 | XP_011525479.1 | 20, 21 | 2449 | |||
XM_011527178.2 | XP_011525480.1 | 21, 22 | 2931 | |||
XM_011527179.1 | XP_011525481.1 | 20, 21 | 2569 | |||
XM_011527182.2 | XP_011525484.1 | 19, 20 | 2313 | |||
XM_017027053.1 | XP_016882542.1 | 19, 20 | 2449 | |||
XM_017027054.1 | XP_016882543.1 | 18, 19 | 2182 | |||
XM_017027055.1 | XP_016882544.1 | 7, 8 | 1658 | |||
GenBank mRNA | AK128220.1 | - | 20, 21 | 2352 | ||
AK310460.1 | - | 16, 17 | 1899 | |||
BC094785.1 | - | 19, 20 | 2628 | |||
BC109035.1 | - | 16, 17 | 1968 | 40028284 |
