siRNA Details
Gene Symbol: ZNF410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242924.1 | NP_001229853.1 | 11 | 1732 | ||
NM_001242927.1 | NP_001229856.1 | 9 | 1462 | |||
NM_001242928.1 | NP_001229857.1 | 10 | 1652 | |||
NM_021188.2 | NP_067011.1 | 10 | 1681 | |||
NR_040251.1 | 10 | 1649 | ||||
GenBank mRNA | AK223261.1 | - | 10 | 1437 | ||
AK293350.1 | - | 11 | 1488 | |||
AK298307.1 | - | 11 | 1404 | |||
AK298520.1 | - | 10 | 1457 | |||
AK299138.1 | - | 9 | 1460 | |||
AK303834.1 | - | 9 | 1141 | |||
AK304609.1 | - | 10 | 1195 | |||
AK309957.1 | - | 11 | 1477 | |||
AK315206.1 | - | 10 | 1439 | |||
BC004357.1 | - | 10 | 1396 | |||
U90919.1 | - | 10 | 1413 |
