siRNA Details
Gene Symbol: PCTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102402.2 | NP_001095872.1 | 2 | 449 | ||
NM_021213.3 | NP_067036.2 | 2 | 342 | |||
XM_005257549.1 | XP_005257606.1 | 2 | 342 | |||
XM_005257550.3 | XP_005257607.1 | 2 | 342 | |||
XM_017024913.1 | XP_016880402.1 | 2 | 424 | |||
GenBank mRNA | AF087857.1 | - | 2 | 327 | ||
AF114430.1 | - | 2 | 232 | |||
AF151638.1 | - | 2 | 326 | |||
AK024667.1 | - | 2 | 268 | |||
AK058120.1 | - | 2 | 275 | |||
AK314509.1 | - | 2 | 268 | |||
BC005112.1 | - | 2 | 154 | |||
BC012084.1 | - | 2 | 260 | 4575824 | ||
BM153142.1 | - | 2 | 342 | |||
BM549250.1 | - | 2 | 449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
