siRNA Details
Gene Symbol: AVPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021732.2 | NP_068378.2 | 3 | 1196 | ||
XM_017016494.1 | XP_016871983.1 | 3 | 1356 | |||
GenBank mRNA | AF131791.1 | - | 3 | 1196 | ||
AF218019.1 | - | 3 | 1151 | |||
AF241786.1 | - | 3 | 1200 | |||
AK223099.1 | - | 3 | 1144 | |||
BC000877.1 | - | 3 | 807 | 3461724 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
