siRNA Details
Gene Symbol: BACH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170794.1 | NP_001164265.1 | 6 | 2557 | ||
NM_021813.3 | NP_068585.1 | 8 | 2728 | |||
XM_005248758.4 | XP_005248815.1 | 5 | 3510 | |||
XM_005248759.4 | XP_005248816.1 | 4 | 3430 | |||
XM_011536039.2 | XP_011534341.1 | 7 | 2639 | |||
XM_011536040.2 | XP_011534342.1 | 6 | 2588 | |||
XM_017011156.1 | XP_016866645.1 | 11 | 3700 | |||
XM_017011157.1 | XP_016866646.1 | 10 | 3506 | |||
XM_017011158.1 | XP_016866647.1 | 12 | 3779 | |||
XM_017011159.1 | XP_016866648.1 | 13 | 3868 | |||
XM_017011160.1 | XP_016866649.1 | 10 | 3622 | |||
XM_017011161.1 | XP_016866650.1 | 12 | 3330 | |||
XM_017011162.1 | XP_016866651.1 | 9 | 3529 | |||
XM_017011163.1 | XP_016866652.1 | 11 | 3701 | |||
XM_017011164.1 | XP_016866653.1 | 10 | 3607 | |||
XM_017011165.1 | XP_016866654.1 | 7 | 2654 | |||
XM_017011166.1 | XP_016866655.1 | 9 | 2812 | |||
XM_017011167.1 | XP_016866656.1 | 11 | 3665 | |||
GenBank mRNA | AB208889.1 | - | 6 | 2194 | ||
AF357835.1 | - | 5 | 2053 | |||
AJ271878.1 | - | 8 | 2562 | |||
HE578166.1 | - | 2 | 408 |
