siRNA Details
Gene Symbol: NIF3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136039.2 | NP_001129511.1 | 2 | 256 | ||
NM_001142355.1 | NP_001135827.1 | 2 | 205 | |||
NM_001142356.1 | NP_001135828.1 | 1 | 191 | |||
NM_021824.3 | NP_068596.2 | 2 | 408 | |||
XM_005246742.3 | XP_005246799.1 | 2 | 471 | |||
XM_011511582.1 | XP_011509884.1 | 2 | 392 | |||
XM_011511583.1 | XP_011509885.1 | 2 | 1136 | |||
XM_011511584.1 | XP_011509886.1 | 2 | 954 | |||
XM_011511585.1 | XP_011509887.1 | 2 | 903 | |||
XM_017004630.1 | XP_016860119.1 | 2 | 653 | |||
GenBank mRNA | AB038949.1 | - | 2 | 369 | ||
AF060513.1 | - | 2 | 165 | |||
AF182416.1 | - | 2 | 327 | |||
AF283538.1 | - | 2 | 211 | |||
AK023378.1 | - | 2 | 354 | |||
AK225905.1 | - | 2 | 1731 | |||
AK314709.1 | - | 2 | 210 | |||
AL834430.1 | - | 1 | 31 | |||
AY251943.1 | - | 1 | 194 | |||
AY251944.1 | - | 1 | 194 | |||
BC007654.2 | - | 1 | 192 | 3346099 | ||
BG724012.1 | - | 2 | 464 | |||
BP194872.1 | - | 2 | 205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
