siRNA Details
Gene Symbol: NIF3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136039.2 | NP_001129511.1 | 5 | 890 | ||
NM_001142355.1 | NP_001135827.1 | 5 | 839 | |||
NM_021824.3 | NP_068596.2 | 5 | 1042 | |||
XM_005246742.3 | XP_005246799.1 | 5 | 1105 | |||
XM_011511582.1 | XP_011509884.1 | 5 | 1026 | |||
XM_011511583.1 | XP_011509885.1 | 5 | 1770 | |||
XM_011511584.1 | XP_011509886.1 | 5 | 1588 | |||
XM_011511585.1 | XP_011509887.1 | 5 | 1537 | |||
XM_017004630.1 | XP_016860119.1 | 5 | 1287 | |||
GenBank mRNA | AB038949.1 | - | 5 | 1003 | ||
AF060513.1 | - | 5 | 799 | |||
AF182416.1 | - | 5 | 964 | |||
AF283538.1 | - | 5 | 845 | |||
AK023378.1 | - | 5 | 988 | |||
AK225905.1 | - | 5 | 2365 | |||
AK314709.1 | - | 5 | 844 | |||
AL834430.1 | - | 4 | 665 | |||
BC007654.2 | - | 4 | 826 | 3346099 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
