siRNA Details
Gene Symbol: NYX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022567.2 | NP_072089.1 | 2 | 2134 | ||
XM_017029709.1 | XP_016885198.1 | 3 | 1802 | |||
GenBank mRNA | AF254868.1 | - | 3 | 1802 | ||
AJ278865.1 | - | 2 | 2160 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
