siRNA Details
Gene Symbol: RIC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286134.1 | NP_001273063.1 | 2 | 1451 | ||
NM_021932.5 | NP_068751.4 | 2 | 1451 | |||
XM_005253052.3 | XP_005253109.1 | 2 | 637 | |||
XM_017018107.1 | XP_016873596.1 | 2 | 721 | |||
GenBank mRNA | AK026897.1 | - | 2 | 1450 | ||
AK027461.1 | - | 2 | 140 | |||
AK225420.1 | - | 2 | 140 | |||
AK314568.1 | - | 2 | 161 | |||
AL136935.1 | - | 2 | 432 | |||
AL390088.1 | - | 2 | 172 | |||
BC111499.1 | - | 2 | 268 | 6526180 | ||
BC121807.1 | - | 2 | 348 | 40108083 | ||
BC121808.1 | - | 2 | 348 | 40108080 | ||
BX538115.1 | - | 1 | 225 |
