siRNA Details
Gene Symbol: PROK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021935.3 | NP_068754.1 | 2, 3 | 369 | ||
GenBank mRNA | AF333025.1 | - | 2, 3 | 224 | ||
BC069395.1 | - | 2, 3 | 224 | 7262718 | ||
BC096695.1 | - | 2, 3 | 224 | 40014094 | ||
BC098110.1 | - | 2, 3 | 224 | 40014092 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
