siRNA Details
Gene Symbol: SLC39A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135146.1 | NP_001128618.1 | 9 | 1707 | ||
NM_001135148.1 | NP_001128620.1 | 8 | 1144 | |||
NM_022154.5 | NP_071437.3 | 8 | 1829 | |||
XM_005263177.1 | XP_005263234.1 | 9 | 1616 | |||
XM_011532182.1 | XP_011530484.1 | 7 | 1642 | |||
XM_017008540.1 | XP_016864029.1 | 9 | 1624 | |||
XM_017008541.1 | XP_016864030.1 | 11 | 2001 | |||
GenBank mRNA | AB020970.1 | - | 3 | 295 | ||
AB040120.1 | - | 8 | 1777 | |||
AF193052.1 | - | 9 | 1658 | |||
AK027652.1 | - | 8 | 1144 | |||
BC001320.1 | - | 2 | 211 | |||
BC012125.1 | - | 9 | 1683 | 4650026 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
