siRNA Details
Gene Symbol: SPATA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310156.1 | NP_001297085.1 | 11, 12 | 1380 | ||
XM_011542514.2 | XP_011540816.1 | 10, 11 | 1254 | |||
GenBank mRNA | AF306347.1 | - | 11, 12 | 1348 |
