siRNA Details
Gene Symbol: AZI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134432.1 | NP_001127904.1 | 3 | 839 | ||
NM_001134433.1 | NP_001127905.1 | 3 | 839 | |||
NM_001271650.1 | NP_001258579.1 | 4 | 903 | |||
NM_022461.4 | NP_071906.1 | 3 | 839 | |||
XM_005265392.2 | XP_005265449.1 | 4 | 926 | |||
XM_011534009.2 | XP_011532311.1 | 3 | 379 | |||
XM_011534010.2 | XP_011532312.1 | 3 | 337 | |||
XM_011534011.2 | XP_011532313.1 | 3 | 336 | |||
GenBank mRNA | AK025592.1 | - | 3 | 686 | ||
AK290637.1 | - | 3 | 558 | |||
AY151386.1 | - | 2 | 307 | |||
BM546093.1 | - | 2 | 175 | |||
BX386357.2 | - | 3 | 760 | |||
BX648471.1 | - | 4 | 567 | |||
CB995808.1 | - | 3 | 367 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
