siRNA Details
Gene Symbol: AZI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271650.1 | NP_001258579.1 | 8 | 1350 | ||
NM_022461.4 | NP_071906.1 | 7 | 1286 | |||
XM_005265392.2 | XP_005265449.1 | 8 | 1373 | |||
XM_011534009.2 | XP_011532311.1 | 7 | 826 | |||
GenBank mRNA | AK025592.1 | - | 7 | 1133 | ||
AY151386.1 | - | 6 | 754 | |||
BM546093.1 | - | 6 | 622 | |||
BX648471.1 | - | 8 | 1014 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
