siRNA Details
Gene Symbol: INF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031714.3 | NP_001026884.3 | 8 | 1830 | ||
NM_022489.3 | NP_071934.3 | 8 | 1830 | |||
XM_005268004.4 | XP_005268061.1 | 8 | 1917 | |||
XM_005268005.4 | XP_005268062.1 | 8 | 1888 | |||
XM_017021595.1 | XP_016877084.1 | 8 | 1888 | |||
GenBank mRNA | BG335490.1 | - | 1 | 130 | ||
DQ395338.1 | - | 1 | 91 | |||
DQ395339.1 | - | 1 | 91 | |||
DQ395340.1 | - | 1 | 91 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
