siRNA Details
Gene Symbol: MTHFSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159377.1 | NP_001152849.1 | 3 | 248 | ||
NM_001159380.1 | NP_001152852.1 | 3 | 212 | |||
NM_022764.2 | NP_073601.2 | 3 | 245 | |||
NR_027490.1 | 3 | 248 | ||||
XM_006721247.3 | XP_006721310.1 | 2 | 119 | |||
XM_011523284.2 | XP_011521586.1 | 3 | 248 | |||
XM_011523286.1 | XP_011521588.1 | 3 | 309 | |||
XM_011523287.1 | XP_011521589.1 | 3 | 213 | |||
XM_011523288.1 | XP_011521590.1 | 2 | 141 | |||
XM_011523289.1 | XP_011521591.1 | 3 | 309 | |||
XM_017023573.1 | XP_016879062.1 | 3 | 202 | |||
GenBank mRNA | AK023060.1 | - | 3 | 223 | ||
AK023955.1 | - | 3 | 202 | |||
BC013911.2 | - | 2 | 179 | |||
BC112057.1 | - | 3 | 228 | 8327525 | ||
BC112059.1 | - | 3 | 228 | 8327527 | ||
BC143704.1 | - | 3 | 213 | |||
BC143705.1 | - | 3 | 212 |
